Giemsa's stain for clinical diagnosis

Supplier: PanReac AppliChem
Danger

251338.1610
APPC251338.1610EA 109 EUR
APPC251338.1610
Giemsa's stain for clinical diagnosis
Giemsa's stain

Azur-eosin-methylene blue solution according to Giemsa (slow) for clinical diagnosis.


Suitable for hematology, blood smear staining.


COMPOSITION:
Azur-Eosin-Methylene Blue dye according to Giemsa: 0.5 g
Methanol: 50 ml
Glycerol: 50 ml

Storage Temperature: Ambient
MDL Number: MFCD00081642
CAS Number: 51811-82-6
UN: 1230
ADR: 3(6.1),II

Order Now

Specification Test Results

Suitability for hematology stain passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR