Anti-ROM1 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
ARP44260_P050
AVIVARP44260_P050EA 376 EUR
AVIVARP44260_P050
Anti-ROM1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ROM1 (retinal outer segment membrane protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR