Water, Filter sterile

Supplier: VWR Chemicals

Synonyms: Hydrogen oxide

AX021403EA 141 EUR
AX021403
Water, Filter sterile
Water
Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR