MultiRack racked tips, Axygen®
Supplier: Corning
MR-200-R MR-200-R-S MR-1000XT-R MR-1000XT-R-S MR-10XT-R MR-10XT-R-S MR-300-R MR-300-R-S MR-300XT-R MR-300XT-R-S
AXYGMR-200-REA 368 EUR
AXYGMR-200-R AXYGMR-200-R-S AXYGMR-1000XT-R AXYGMR-1000XT-R-S AXYGMR-10XT-R AXYGMR-10XT-R-S AXYGMR-300-R AXYGMR-300-R-S AXYGMR-300XT-R AXYGMR-300XT-R-S
MultiRack racked tips, Axygen®
Pipette Tips
The Axygen® MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in bulk, racked, filter and reload packaging styles to meet the variety of needs in the lab setting.
  • Detectable DNase/RNase/Human DNA/PCR inhibition free
  • Non pyrogenic
  • Manufactured in an ISO certified facility in the USA
  • Fully autoclavable
Axygen® MultiRack tips are supplied in convenient hinged racks that help reduce airborne contamination of the tips during use. Each rack is individually sealed. The high-More Product Information