Sedimentation cones, Imhoff type, Azlon®
Supplier: SciLabware
SWX008
AZLOSWX008EA 207 EUR
AZLOSWX008
Sedimentation cones, Imhoff type, Azlon®
Funnels Sedimentation Cones
Colourless cone, made up of transparent styrene-acrylonitrile S.A.N polymer.
  • With moulded graduations
  • Stopper at the bottom facilitates the removal of sample and cleaning
Certifications: Complies to DIN 12672