N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane, Cerilliant® for GC derivatisation, analytical standard, Supelco®

Supplier: Merck
Danger

B-023-10X1ML
B-023-10X1MLEA 115 EUR
B-023-10X1ML
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane, Cerilliant® for GC derivatisation, analytical standard, Supelco®
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane
Formula: C₈H₁₈F₃NOSi₂
Storage Temperature: Freezer
MDL Number: MFCD00008269
CAS Number: 25561-30-2
UN: 2920
ADR: 8(3),II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR