Ethyl-2-oxo-4-phenylbutyrate ≥90%

Supplier: Thermo Fisher Scientific

B20148.09 B20148.18
B20148.09EA 82.4 EUR
B20148.09 B20148.18
Ethyl-2-oxo-4-phenylbutyrate ≥90%
Ethyl-2-oxo-4-phenylbutyrate

Purity: 90+% CAS No.: 64920-29-2 Molecular Formula: C12H14O3 Formula Weight: 206.24 Flash Point: 110°(230°F) Refractive Index: 1.5020 Boiling Point: 131-133°/2mm Beilstein Reference: 2725083 MDL No.: MFCD00037533

Formula: C₁₂H₁₄O₃
Boiling Pt: 131…133 °C (2 mmHg)
Density: 1,09
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00037533
CAS Number: 64920-29-2
EINECS: 265-276-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR