14-Heptacosanone ≥97%

Supplier: Thermo Fisher Scientific

B20191.06 B20191.14
B20191.06EA 78.1 EUR
B20191.06 B20191.14
14-Heptacosanone ≥97%
14-Heptacosanone

Purity: 97% CAS No.: 542-50-7 Molecular Formula: C27H54O Formula Weight: 394.73 Melting Point: 76-79° Beilstein Reference: 1796794

Formula: C₂₇H₅₄O
Melting Pt: 76…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00026598
CAS Number: 542-50-7
EINECS: 208-815-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR