3,5-Bis(trifluoromethyl)phenyl isothiocyanate ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 1,3-Bis(trifluoromethyl)-5-isothiocyanatobenzene, 1-Isothiocyanato-3,5-bis(trifluoromethyl)benzene

B20206.03 B20206.06 B20206.14
B20206.03EA 36.6 EUR
B20206.03 B20206.06 B20206.14
3,5-Bis(trifluoromethyl)phenyl isothiocyanate ≥98%
3,5-Bis(trifluoromethyl)phenyl isothiocyanate

Purity: 98% CAS No.: 23165-29-9 Molecular Formula: C9H3F6NS Formula Weight: 271.19 Flash Point: 108°(226°F) Refractive Index: 1.500 Odour: Irritating Boiling Point: 109-110°/35mm Beilstein Reference: 2990816 UN No.: 2922 MDL No.: MFCD00040838

Formula: C₉H₃F₆NS
MW: 271.19 g/mol
Boiling Pt: 109…110 °C (35 mmHg)
Density: 1.485
Flash Pt: 108 °C (226 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040838
CAS Number: 23165-29-9
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR