2-Chloro-4-fluorotoluene ≥98%

Supplier: Thermo Fisher Scientific
Warning

B20278.14 B20278.22 B20278.36
B20278.14EA 34 EUR
B20278.14 B20278.22 B20278.36
2-Chloro-4-fluorotoluene ≥98%
2-Chloro-4-fluorotoluene

Purity: 98% CAS No.: 452-73-3 Molecular Formula: C7H6ClF Formula Weight: 144.58 Flash Point: 50°(122°F) Refractive Index: 1.4990 Odour: Aromatic Boiling Point: 154-156° Beilstein Reference: 1931690 UN No.: 1993 MDL No.: MFCD00000572

Formula: C₇H₆ClF
Boiling Pt: 154…156 °C
Density: 1,198
Flash Pt: 50 °C (122 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000572
CAS Number: 452-73-3
EINECS: 207-209-8
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR