5-Chloro-2-thiophenesulphonamide 97%

Supplier: Thermo Fisher Scientific
Warning

B20477.03 B20477.06
B20477.03EA 15.9 EUR
B20477.03 B20477.06
5-Chloro-2-thiophenesulphonamide 97%
5-Chloro-2-thiophenesulphonamide

Purity: 97% CAS No.: 53595-66-7 Molecular Formula: C4H4ClNO2S2 Formula Weight: 197.67 Melting Point: 115-118° MDL No.: MFCD00052584

Formula: C₄H₄ClNO₂S₂
Melting Pt: 115…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00052584
CAS Number: 53595-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR