5-Methyl-1-hexene ≥99%

Supplier: Thermo Fisher Scientific

B20719.14
B20719.14EA 0 EUR
B20719.14
5-Methyl-1-hexene ≥99%
5-Methyl-1-hexene

Purity: 99% CAS No.: 3524-73-0 Molecular Formula: (CH3)2CHCH2CH2CH=CH2 Formula Weight: 98.19 Flash Point: -10°(14°F) Refractive Index: 1.396 Boiling Point: 85° Beilstein Reference: 1731846 UN No.: 3295 MDL No.: MFCD00039858

Formula: C₇H₁₄
MW: 98.19 g/mol
Boiling Pt: 85
Melting Pt: –122 °C
Density: 0.692 g/cm³ (20 °C)
Flash Pt: –10 °C (14 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039858
CAS Number: 3524-73-0
EINECS: 222-541-3
UN: 3295
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR