2-Acetyl-3-amino-5-phenylthiophene 97%

Supplier: Thermo Fisher Scientific
Warning

B20751.02
B20751.02EA 0 EUR
B20751.02
2-Acetyl-3-amino-5-phenylthiophene 97%
2-Acetyl-3-amino-5-phenylthiophene

Purity: 97% CAS No.: 105707-24-2 Molecular Formula: C12H11NOS Formula Weight: 217.29 Melting Point: 119-122° MDL No.: MFCD00068132

Formula: C₁₂H₁₁NOS
MW: 217.29 g/mol
Melting Pt: 119…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00068132
CAS Number: 105707-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR