2,4-Dichlorobenzoyl chloride ≥98%

Supplier: Thermo Fisher Scientific
Danger

B20815.18 B20815.30
B20815.18EA 0 EUR
B20815.18 B20815.30
2,4-Dichlorobenzoyl chloride ≥98%
2,4-Dichlorobenzoyl chloride

Purity: 98% CAS No.: 89-75-8 Molecular Formula: C7H3Cl3O Formula Weight: 209.46 Flash Point: 137°(278°F) Refractive Index: 1.5930 Melting Point: 16-20° Boiling Point: 150°/34mm Beilstein Reference: 608234 UN No.: 3265 MDL No.: MFCD00000661

Formula: C₇H₃Cl₃O
Boiling Pt: 150 °C (34 mmHg)
Melting Pt: 16…20 °C
Density: 1,497
Flash Pt: 137 °C (278 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000661
CAS Number: 89-75-8
EINECS: 201-936-4
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR