L(+)-Ribose ≥99%

Supplier: Thermo Fisher Scientific

Synonyms: (2S,3S,4S)-2,3,4,5-Tetrahydroxypentanal, L-(+)-Ribose

B21117.03 B21117.06 B21117.14
B21117.03EA 102 EUR
B21117.03 B21117.06 B21117.14
L(+)-Ribose ≥99%
L(+)-Ribose

Purity: 99% CAS No.: 24259-59-4 Molecular Formula: C5H10O5 Formula Weight: 150.13 Melting Point: 85-88° MDL No.: MFCD00167010

Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Boiling Pt: 331
Melting Pt: 85…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00167010
CAS Number: 24259-59-4
EINECS: 246-110-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR