3-Chloro-o-xylene ≥97%

Supplier: Thermo Fisher Scientific
Warning

B21505.14 B21505.22
B21505.14EA 79.3 EUR
B21505.14 B21505.22
3-Chloro-o-xylene ≥97%
3-Chloro-o-xylene

Purity: 97% CAS No.: 608-23-1 Molecular Formula: C8H9Cl Formula Weight: 140.61 MDL No.: MFCD00060689

Formula: C₈H₉Cl
Storage Temperature: Ambient
MDL Number: MFCD00060689
CAS Number: 608-23-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR