1,2-Bis(tosyloxy)ethane 97%

Supplier: Thermo Fisher Scientific
Warning

B21539.09 B21539.18 B21539.30
B21539.09EA 66.2 EUR
B21539.09 B21539.18 B21539.30
1,2-Bis(tosyloxy)ethane 97%
1,2-Bis(tosyloxy)ethane

Purity: 97% CAS No.: 6315-52-2 Molecular Formula: C16H18O6S2 Formula Weight: 370.45 Melting Point: 122-127° MDL No.: MFCD00008549

Formula: C₁₆H₁₈O₆S₂
Melting Pt: 122…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00008549
CAS Number: 6315-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR