15-Hydroxypentadecanoic acid ≥99%

Supplier: Thermo Fisher Scientific

B21670.03 B21670.06
B21670.03EA 62.8 EUR
B21670.03 B21670.06
15-Hydroxypentadecanoic acid ≥99%
15-Hydroxypentadecanoic acid

Purity: 99+% CAS No.: 4617-33-8 Molecular Formula: C15H30O3 Formula Weight: 258.40 Melting Point: 84-88° Beilstein Reference: 1240100

Formula: C₁₅H₃₀O₃
Melting Pt: 84…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00039535
CAS Number: 4617-33-8
EINECS: 225-026-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR