TES (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid) ≥99%

Supplier: Thermo Fisher Scientific
Warning

B21819.09 B21819.18 B21819.30
B21819.09EA 39.6 EUR
B21819.09 B21819.18 B21819.30
TES (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid) ≥99%
TES (2-(Tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid)

Purity: 99% CAS No.: 7365-44-8 Molecular Formula: C6H15NO6S Formula Weight: 229.25 Odour: Odourless Melting Point: ca 225° dec. Beilstein Reference: 1957061 Merck Reference: 14,9176 MDL No.: MFCD00007532

Formula: C₆H₁₅NO₆S
Melting Pt: ∼ 225 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007532
CAS Number: 7365-44-8
EINECS: 230-906-3
Merck Index: 14,9176

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR