4-Amino-2-chloropyridine 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Chloro-4-aminopyridine

B22098.06 B22098.14
B22098.06EA 73.1 EUR
B22098.06 B22098.14
4-Amino-2-chloropyridine 98%
4-Amino-2-chloropyridine

Purity: 98% CAS No.: 14432-12-3 Molecular Formula: C5H5ClN2 Formula Weight: 128.56 Odour: Unpleasant Melting Point: 91-94° Boiling Point: 153°/5mm Beilstein Reference: 108671 MDL No.: MFCD00060089

Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Boiling Pt: 153 °C (5 mmHg)
Melting Pt: 91…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00060089
CAS Number: 14432-12-3
EINECS: 238-403-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR