2-Ethyl-4,4-dimethyl-2-oxazoline ≥97%

Supplier: Thermo Fisher Scientific

B22117.14
B22117.14EA 0 EUR
B22117.14
2-Ethyl-4,4-dimethyl-2-oxazoline ≥97%
2-Ethyl-4,4-dimethyl-2-oxazoline

Purity: 97% CAS No.: 5146-88-3 Molecular Formula: C7H13NO Formula Weight: 127.19 Boiling Point: 129-130° UN No.: 1993 MDL No.: MFCD00014506

Formula: C₇H₁₃NO
Boiling Pt: 129…130 °C
MDL Number: MFCD00014506
CAS Number: 5146-88-3
EINECS: 225-919-6
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR