(4-Bromophenyl)acetic acid ethyl ester ≥98%

Supplier: Thermo Fisher Scientific
Warning

B22165.14 B22165.06 B22165.22
B22165.14EA 178 EUR
B22165.14 B22165.06 B22165.22
(4-Bromophenyl)acetic acid ethyl ester ≥98%
(4-Bromophenyl)acetic acid ethyl ester

Purity: 98% CAS No.: 14062-25-0 Molecular Formula: C10H11BrO2 Formula Weight: 243.10 Melting Point: 29-31° Boiling Point: 88-90°/0.35mm MDL No.: MFCD00016333

Formula: C₁₀H₁₁BrO₂
Boiling Pt: 88…90 °C (0,35 mmHg)
Melting Pt: 29…31 °C
Storage Temperature: Ambient
MDL Number: MFCD00016333
CAS Number: 14062-25-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR