3-Chloro-2-methylanisole ≥97%

Supplier: Thermo Fisher Scientific
Warning

B22203.06 B22203.14 B22203.22
B22203.06EA 59.9 EUR
B22203.06 B22203.14 B22203.22
3-Chloro-2-methylanisole ≥97%
3-Chloro-2-methylanisole

Purity: 97% CAS No.: 3260-88-6 Molecular Formula: C8H9ClO Formula Weight: 156.61 Flash Point: 92°(197°F) Refractive Index: 1.5395 Melting Point: -3° Boiling Point: 212-215° Beilstein Reference: 1862220 UN No.: 1993 MDL No.: MFCD00070772

Formula: C₈H₉ClO
Boiling Pt: 212…215 °C
Melting Pt: –3 °C
Density: 1,160
Flash Pt: 92 °C (197 °F)
Storage Temperature: Ambient
MDL Number: MFCD00070772
CAS Number: 3260-88-6
EINECS: 221-862-6
ADR: 3,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR