Fuchsin (acid)

Supplier: Thermo Fisher Scientific
Warning

B22222.14 B22222.22 B22222.36
B22222.14EA 87.7 EUR
B22222.14 B22222.22 B22222.36
Fuchsin (acid)
Fuchsin (acid)

CAS No.: 3244-88-0 Molecular Formula: C20H17N3Na2O9S3 Formula Weight: 585.53 Beilstein Reference: 4111656 Merck Reference: 14,107 MDL No.: MFCD00013286

Formula: C₂₀H₁₇N₃Na₂O₉S₃
Melting Pt: > 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00013286
CAS Number: 3244-88-0
EINECS: 221-816-5
Merck Index: 14,107

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR