4-Chloro-m-toluidine ≥98%

Supplier: Thermo Fisher Scientific
Danger

B22234.06 B22234.14
B22234.06EA 68.5 EUR
B22234.06 B22234.14
4-Chloro-m-toluidine ≥98%
4-Chloro-m-toluidine

Purity: 98% CAS No.: 7149-75-9 Molecular Formula: C7H8ClN Formula Weight: 141.60 Melting Point: 82-85° Beilstein Reference: 636522 UN No.: 2239 MDL No.: MFCD00066332

Formula: C₇H₈ClN
Boiling Pt: 241 °C (1013 hPa)
Melting Pt: 82…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00066332
CAS Number: 7149-75-9
UN: 2239
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR