(R)-(-)-2-Methylpiperazine ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: (R)-2-Methylpiperazine

B22366.01
B22366.01EA 0 EUR
B22366.01
(R)-(-)-2-Methylpiperazine ≥98%
(R)-(-)-2-Methylpiperazine

Purity: 98+% CAS No.: 75336-86-6 Molecular Formula: C5H12N2 Formula Weight: 100.17 Flash Point: 65°(149°F) Melting Point: 76-78° Boiling Point: 155-156° Beilstein Reference: 3647994 Optical Rotation: -7° (c=1 in ethanol) UN No.: 2925 MDL No.: MFCD00192317

Formula: C₅H₁₂N₂
MW: 100.16 g/mol
Boiling Pt: 155…156 °C
Melting Pt: 76…78 °C
Flash Pt: 65 °C (149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00192317
CAS Number: 75336-86-6
UN: 2925
ADR: 4.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR