4-(Trifluoromethyl)phenylboronic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

B22374.03 B22374.06 B22374.14
B22374.03EA 53.9 EUR
B22374.03 B22374.06 B22374.14
4-(Trifluoromethyl)phenylboronic acid ≥98%
4-(Trifluoromethyl)phenylboronic acid

Purity: 98% CAS No.: 128796-39-4 Molecular Formula: C7H6BF3O2 Formula Weight: 189.93 Melting Point: 238-242° Beilstein Reference: 3544189

Formula: C₇H₆BF₃O₂
Melting Pt: 238…242 °C
Storage Temperature: Ambient
MDL Number: MFCD00151855
CAS Number: 128796-39-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR