tert-Butyl methyl sulphide ≥98%

Supplier: Thermo Fisher Scientific
Danger

B22390.06 B22390.14
B22390.06EA 41.5 EUR
B22390.06 B22390.14
tert-Butyl methyl sulphide ≥98%
tert-Butyl methyl sulphide

Purity: 98+% CAS No.: 6163-64-0 Molecular Formula: C5H12S Formula Weight: 104.22 Flash Point: -3°(26°F) Refractive Index: 1.4400 Odour: Unpleasant Boiling Point: 101-102° UN No.: UN1993 MDL No.: MFCD00008840

Formula: C₅H₁₂S
Boiling Pt: 101…102 °C
Density: 0,826
Flash Pt: –3 °C (26 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008840
CAS Number: 6163-64-0
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR