Ethylphosphonic acid ≥98%

Supplier: Thermo Fisher Scientific
Danger

B22559.03 B22559.06 B22559.14
B22559.03EA 43.4 EUR
B22559.03 B22559.06 B22559.14
Ethylphosphonic acid ≥98%
Ethylphosphonic acid

Purity: 98+% CAS No.: 6779-09-5 Molecular Formula: C2H7O3P Formula Weight: 110.06 Melting Point: 58-62° Beilstein Reference: 1743032 UN No.: 3261 MDL No.: MFCD00002141

Formula: C₂H₇O₃P
MW: 110.05 g/mol
Melting Pt: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00002141
CAS Number: 6779-09-5
EINECS: 229-839-2
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR