3',5'-Dimethoxy-4'-hydroxyacetophenone ≥97%

Supplier: Thermo Fisher Scientific
Warning

B22634.03 B22634.06 B22634.14
B22634.03EA 52.4 EUR
B22634.03 B22634.06 B22634.14
3',5'-Dimethoxy-4'-hydroxyacetophenone ≥97%
3',5'-Dimethoxy-4'-hydroxyacetophenone

Purity: 97% CAS No.: 2478-38-8 Molecular Formula: C10H12O4 Formula Weight: 196.2 Melting Point: 117-122° Beilstein Reference: 1966119

Formula: C₁₀H₁₂O₄
Boiling Pt: 335 °C (1013 hPa)
Melting Pt: 117…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00008748
CAS Number: 2478-38-8
EINECS: 219-610-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR