Dimethyl suberate ≥99%

Supplier: Thermo Fisher Scientific

B22681.14 B22681.22
B22681.14EA 59.4 EUR
B22681.14 B22681.22
Dimethyl suberate ≥99%
Dimethyl suberate

Purity: 99% CAS No.: 1732-09-8 Molecular Formula: CH3O2C(CH2)6CO2CH3 Formula Weight: 202.25 Flash Point: 110°(230°F) Refractive Index: 1.4320 Boiling Point: 266-268° Beilstein Reference: 1780054 MDL No.: MFCD00008471

Formula: C₁₀H₁₈O₄
Boiling Pt: 266…268 °C
Melting Pt: –3,1 °C
Density: 1,016
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008471
CAS Number: 1732-09-8
EINECS: 257-059-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR