Boc-D-Alanine ≥98%

Supplier: Thermo Fisher Scientific

Synonyms: N-Boc-D-alanine, Boc-D-Ala-OH

B22706.06 B22706.14
B22706.06EA 46.1 EUR
B22706.06 B22706.14
Boc-D-Alanine ≥98%
Boc-D-Alanine

Purity: 98+% CAS No.: 7764-95-6 Molecular Formula: C8H15NO4 Formula Weight: 189.21 Melting Point: 81-84° Beilstein Reference: 2048396 Optical Rotation: +26° (c=2 in acetic acid) MDL No.: MFCD00063123

Formula: C₈H₁₅NO₄
MW: 189.2 g/mol
Melting Pt: 81…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00063123
CAS Number: 7764-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR