Diethyl pyrocarbonate ≥97%

Supplier: Thermo Fisher Scientific
Warning

B22753.06 B22753.14 B22753.22
B22753.06EA 31.7 EUR
B22753.06 B22753.14 B22753.22
Diethyl pyrocarbonate ≥97%
Diethyl pyrocarbonate

Purity: 97% CAS No.: 1609-47-8 Molecular Formula: C6H10O5 Formula Weight: 162.14 Storage Temperature: 2°C to 8°C Flash Point: 69°(156°F) Physical Form: Clear liquid Density: 1.101 Refractive Index: 1.3980 Sensitivity: Moisture Sensitive Appearance: Colourless Boiling Point: 93-94°/18mm Beilstein Reference: 637031 Merck Reference: 14,7998 MDL No.: MFCD00009106


Ribonuclease inhibitor

Formula: O(COOC₂H₅)₂
Boiling Pt: 93…94 °C (18 mmHg)
Density: 1,101
Flash Pt: 69 °C (156 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00009106
CAS Number: 1609-47-8
EINECS: 216-542-8
Merck Index: 14,7998

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR