3,5-Dichlorophenylboronic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

B22765.03 B22765.06
B22765.03EA 50.2 EUR
B22765.03 B22765.06
3,5-Dichlorophenylboronic acid ≥98%
3,5-Dichlorophenylboronic acid

Purity: 98+% CAS No.: 67492-50-6 Molecular Formula: C6H5BCl2O2 Formula Weight: 190.82 Melting Point: ca 315° dec. Beilstein Reference: 7581091

Formula: C₆H₅BCl₂O₂
Melting Pt: ∼ 315 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00051935
CAS Number: 67492-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR