3,4-Difluorophenylboronic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 3,4-Difluorobenzeneboronic acid, (3,4-Difluorophenyl)metaboric acid, (3,4-Difluorophenyl)boronic acid, Dihydroxy(3,4-difluorophenyl)borane

B22799.03 B22799.06 B22799.14
B22799.03EA 15.2 EUR
B22799.03 B22799.06 B22799.14
3,4-Difluorophenylboronic acid ≥98%
3,4-Difluorophenylboronic acid

Purity: 98% CAS No.: 168267-41-2 Molecular Formula: C6H5BF2O2 Formula Weight: 157.91 Melting Point: 300° Beilstein Reference: 7369788

Formula: C₆H₅BF₂O₂
MW: 157.91 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00807405
CAS Number: 168267-41-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR