Benzyltrimethylammonium dichloroiodate 95%

Supplier: Thermo Fisher Scientific
Warning

B22892.06 B22892.14
B22892.06EA 58.2 EUR
B22892.06 B22892.14
Benzyltrimethylammonium dichloroiodate 95%
Benzyltrimethylammonium dichloroiodate

Purity: 95% CAS No.: 114971-52-7 Molecular Formula: C10H16NICl2 Formula Weight: 348.06 Melting Point: ca. 125° Beilstein Reference: 4611949

Formula: C₁₀H₁₆Cl₂IN
Melting Pt: ∼, 125 °C
Storage Temperature: Ambient
MDL Number: MFCD00075259
CAS Number: 114971-52-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR