Dipentyl phthalate ≥98%

Supplier: Thermo Fisher Scientific
Danger

B22910.06 B22910.14 B22910.22
B22910.06EA 95.4 EUR
B22910.06 B22910.14 B22910.22
Dipentyl phthalate ≥98%
Dipentyl phthalate

Purity: 98% CAS No.: 131-18-0 Molecular Formula: C18H26O4 Formula Weight: 306.41 Flash Point: 118°(244°F) Odour: Odourless Boiling Point: 342° UN No.: 3082 MDL No.: MFCD00041934

Formula: C₁₈H₂₆O₄
Boiling Pt: 342 °C
Density: 1,025
Flash Pt: 118 °C (244 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041934
CAS Number: 131-18-0
EINECS: 205-017-9
UN: 3082
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR