(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (2R)-(+)-2-Methoxy-2-phenyl-3,3,3-trifluoropropanoic acid, (R)-3,3,3-Trifluoro-2-methoxy-2-phenylpropanoic acid

B22968.77 B22968.88 B22968.ME
B22968.77EA 36.6 EUR
B22968.77 B22968.88 B22968.ME
(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid ≥99%
(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid

Purity: 99% CAS No.: 20445-31-2 Molecular Formula: C10H9F3O3 Formula Weight: 234.18 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.4730 Melting Point: 43-45° Boiling Point: 105-107°/1mm Beilstein Reference: 2215440 Merck Reference: 14,6280 Optical Rotation: +72° (c=2 in methanol) MDL No.: MFCD00004184

Formula: C₁₀H₉F₃O₃
MW: 234.17 g/mol
Boiling Pt: 105…107 °C (1 mmHg)
Melting Pt: 43…45 °C
Density: 1.344
Flash Pt: >110 °C (230 °F)
MDL Number: MFCD00004184
CAS Number: 20445-31-2
EINECS: 243-829-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR