Formaldehyde diethyl acetal ≥99% stabilised

Supplier: Thermo Fisher Scientific
Danger


Stabilisation: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

B23134.AE B23134.AP
B23134.AEEA 53.9 EUR
B23134.AE B23134.AP
Formaldehyde diethyl acetal ≥99% stabilised
Formaldehyde diethyl acetal

Stabilized with 100 ppm BHT


Purity: 99% CAS No.: 462-95-3 Molecular Formula: CH3CH2OCH2OCH2CH3 Formula Weight: 104.15 Flash Point: -5°(23°F) Physical Form: Liquid Density: 0.831 Refractive Index: 1.3740 Melting Point: -66° Boiling Point: 87-88° Beilstein Reference: 1697253 UN No.: 2373 MDL No.: MFCD00009246

Formula: C₅H₁₂O₂
Boiling Pt: 87…88 °C
Melting Pt: –66 °C
Density: 0,831
Flash Pt: –5 °C (23 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009246
CAS Number: 462-95-3
EINECS: 207-330-6
UN: 2373
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR