1,8-Dichloro-9,10-anthraquinone 96%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1,8-Dichloroanthraquinone, 1,8-Dichloroanthracene-9,10-dione

B23421.14 B23421.22 B23421.36
B23421.14EA 48.4 EUR
B23421.14 B23421.22 B23421.36
1,8-Dichloro-9,10-anthraquinone 96%
1,8-Dichloro-9,10-anthraquinone

Purity: 96% CAS No.: 82-43-9 Molecular Formula: C14H6Cl2O2 Formula Weight: 277.11 Melting Point: 201-203° MDL No.: MFCD00001191

Formula: C₁₄H₆Cl₂O₂
MW: 277.11 g/mol
Melting Pt: 201…203 °C
Storage Temperature: Ambient
MDL Number: MFCD00001191
CAS Number: 82-43-9
EINECS: 201-420-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR