Benzyltrimethylammonium iodide ≥98%

Supplier: Thermo Fisher Scientific

B23520.06 B23520.22
B23520.06EA 0 EUR
B23520.06 B23520.22
Benzyltrimethylammonium iodide ≥98%
Benzyltrimethylammonium iodide

Purity: 98% CAS No.: 4525-46-6 Molecular Formula: C6H5CH2N(CH3)3I Formula Weight: 277.15 Melting Point: 177-182° Beilstein Reference: 3917007

Formula: C₁₀H₁₆IN
Melting Pt: 179 °C
MDL Number: MFCD00051859
CAS Number: 4525-46-6
EINECS: 224-861-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR