N,N'-Diphenyl-p-phenylenediamine 97%

Supplier: Thermo Fisher Scientific
Warning

B23713.06 B23713.14 B23713.22
B23713.06EA 36.9 EUR
B23713.06 B23713.14 B23713.22
N,N'-Diphenyl-p-phenylenediamine 97%
N,N'-Diphenyl-p-phenylenediamine

Purity: 97% CAS No.: 74-31-7 Molecular Formula: C18H16N2 Formula Weight: 260.34 Melting Point: 146-153° Boiling Point: 220-225°/1mm Merck Reference: 14,3329 MDL No.: MFCD00003015

Formula: C₁₈H₁₆N₂
Boiling Pt: 220…225 °C (1 mmHg)
Melting Pt: 146…153 °C
Density: 1,28
Storage Temperature: Ambient
MDL Number: MFCD00003015
CAS Number: 74-31-7
EINECS: 200-806-4
Merck Index: 14,3329

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR