4-Fluoro-2-methylphenylboronic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

B24117.06 B24117.03 B24117.14
B24117.06EA 195 EUR
B24117.06 B24117.03 B24117.14
4-Fluoro-2-methylphenylboronic acid ≥98%
4-Fluoro-2-methylphenylboronic acid

Purity: 98% CAS No.: 139911-29-8 Molecular Formula: C7H8BFO2 Formula Weight: 153.95 Melting Point: 199-201° MDL No.: MFCD02093072

Formula: C₇H₈BFO₂
Melting Pt: 199…201 °C
Storage Temperature: Ambient
MDL Number: MFCD02093072
CAS Number: 139911-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR