4-Bromo-2,2-difluoro-1,3-benzodioxole ≥97%

Supplier: Thermo Fisher Scientific
Warning

B24389.03 B24389.06
B24389.03EA 63.3 EUR
B24389.03 B24389.06
4-Bromo-2,2-difluoro-1,3-benzodioxole ≥97%
4-Bromo-2,2-difluoro-1,3-benzodioxole

Purity: 97% CAS No.: 144584-66-7 Molecular Formula: C7H3BrF2O2 Formula Weight: 237.00 Refractive Index: 1.4968

Formula: C₇H₃BrF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD01631385
CAS Number: 144584-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR