β-D-Ribofuranose-1,2,3,5-tetraacetate ≥98%

Supplier: Thermo Fisher Scientific

B24426.06 B24426.14 B24426.22
B24426.06EA 43.5 EUR
B24426.06 B24426.14 B24426.22
β-D-Ribofuranose-1,2,3,5-tetraacetate ≥98%
beta-D-Ribofuranose-1,2,3,5-tetraacetate

Purity: 98+% CAS No.: 13035-61-5 Molecular Formula: C13H18O9 Formula Weight: 318.28 Melting Point: 80-83° Beilstein Reference: 94078 Optical Rotation: -13+/-0.5° (c=2.4 in chloroform) MDL No.: MFCD00005358

Formula: C₁₃H₁₈O₉
Melting Pt: 80…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00005358
CAS Number: 13035-61-5
EINECS: 235-898-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR