3-Chlorophenylboronic acid ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 3-Chlorobenzeneboronic acid, (3-Chlorophenyl)metaboric acid, 3-Chlorophenyl boronic acid, Dihydroxy(3-chlorophenyl)borane

B24444.06 B24444.03 B24444.14
B24444.06EA 75.2 EUR
B24444.06 B24444.03 B24444.14
3-Chlorophenylboronic acid ≥97%
3-Chlorophenylboronic acid

Purity: 97% CAS No.: 63503-60-6 Molecular Formula: C6H6BClO2 Formula Weight: 156.38 Melting Point: 180-185° Beilstein Reference: 2936343

Formula: C₆H₆BClO₂
MW: 156.38 g/mol
Melting Pt: 180…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00161354
CAS Number: 63503-60-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR