2,6-Dichlorotoluene ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1,3-Dichloro-2-methylbenzene

B24463.14 B24463.22 B24463.36
B24463.14EA 29.9 EUR
B24463.14 B24463.22 B24463.36
2,6-Dichlorotoluene ≥99%
2,6-Dichlorotoluene

Purity: 99% CAS No.: 118-69-4 Molecular Formula: C7H6Cl2 Formula Weight: 161.03 Flash Point: 82°(179°F) Refractive Index: 1.5510 Melting Point: 2° Boiling Point: 196-203° Beilstein Reference: 1907194 UN No.: 3082 MDL No.: MFCD00000576

Formula: C₇H₆Cl₂
Boiling Pt: 196…203 °C
Melting Pt: 2 °C
Density: 1,254
Flash Pt: 82 °C (179 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000576
CAS Number: 118-69-4
EINECS: 204-269-7
UN: 3082
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR