1-(2-Aminoethyl)pyrrolidine 99%

Supplier: Thermo Fisher Scientific
Danger

B24645.06 B24645.14 B24645.22
B24645.06EA 58.7 EUR
B24645.06 B24645.14 B24645.22
1-(2-Aminoethyl)pyrrolidine 99%
1-(2-Aminoethyl)pyrrolidine

Purity: 99% CAS No.: 7154-73-6 Molecular Formula: C6H14N2 Formula Weight: 114.19 Flash Point: 47°(116°F) Refractive Index: 1.4687 Odour: Amine-like Boiling Point: 161° Beilstein Reference: 102978 UN No.: 2734 MDL No.: MFCD00003182

Formula: C₆H₁₄N₂
Boiling Pt: 161 °C
Density: 0,904
Flash Pt: 47 °C (116 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003182
CAS Number: 7154-73-6
EINECS: 230-509-5
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR