Dimidium bromide 95%

Supplier: Thermo Fisher Scientific
Warning

B24818.01 B24818.03
B24818.01EA 85.4 EUR
B24818.01 B24818.03
Dimidium bromide 95%
Dimidium bromide

Purity: 95% CAS No.: 518-67-2 Molecular Formula: C20H18BrN3 Formula Weight: 380.31 Melting Point: 230° dec. MDL No.: MFCD00011757

Formula: C₂₀H₁₈BrN₃
Melting Pt: 230 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011757
CAS Number: 518-67-2
EINECS: 208-256-7
UN: 2811
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e669239c79726ce91f44ef372cbfed426e90f93b&vid=74b03720866852364cb9050de861a4f7c0e5e57b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR