L-Glutamic acid 5-methyl ester ≥99%

Supplier: Thermo Fisher Scientific

B24859.06 B24859.14 B24859.22
B24859.06EA 27.6 EUR
B24859.06 B24859.14 B24859.22
L-Glutamic acid 5-methyl ester ≥99%
L-Glutamic acid 5-methyl ester

Purity: 99% CAS No.: 1499-55-4 Molecular Formula: C6H11NO4 Formula Weight: 161.16 Storage Temperature: 2°C to 8°C Melting Point: 182° dec. MDL No.: MFCD00002632

Formula: C₆H₁₁NO₄
Melting Pt: 182 °C (dec.)
MDL Number: MFCD00002632
CAS Number: 1499-55-4
EINECS: 216-110-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR